Assay Details
| Gene Symbol: SERPINF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078662.1 | NP_001072130.1 | 8 | 1251 | 79 | |
| XM_021066264.1 | XP_020921923.1 | 1364 | 79 | |||
| XM_021066265.1 | XP_020921924.1 | 1272 | 79 | |||
| GenBank mRNA | AK232800.1 | - | 8 | 1324 | 79 | |
| AK346306.1 | - | 8 | 1250 | 79 | ||
| AK399401.1 | - | 5 | 864 | 79 | ||
| DQ789896.1 | - | 8 | 1251 | 79 | ||
More Information
Set Membership: |
|

SERPINF1
Genomic Map