Assay Details
| Gene Symbol: SEPHS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093735.1 | NP_001087204.1 | 1 | 1348 | 79 | |
| GenBank mRNA | EF033624.2 | - | 1 | 1348 | 79 | |
More Information
Set Membership: |
|

SEPHS2
Genomic Map