Assay Details
| Gene Symbol: VDR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097414.1 | NP_001090883.1 | 10 | 1349 | 71 | |
| XM_021091108.1 | XP_020946767.1 | 1527 | 71 | |||
| XM_021091109.1 | XP_020946768.1 | 1663 | 71 | |||
| GenBank mRNA | EF405627.1 | - | 10 | 1349 | 71 | |
More Information
Set Membership: |
|

VDR
Genomic Map