Assay Details
| Gene Symbol: MAVS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097429.1 | NP_001090898.1 | 6 | 1457 | 69 | |
| XM_005672763.3 | XP_005672820.1 | 1633 | 69 | |||
| XM_021077145.1 | XP_020932804.1 | 1568 | 69 | |||
| XM_021077146.1 | XP_020932805.1 | 1740 | 69 | |||
| GenBank mRNA | AB287431.1 | - | 6 | 1457 | 69 | |
| EU082069.1 | - | 7 | 1536 | 69 | ||
| JN644269.1 | - | 6 | 1457 | 69 | ||
| KC860782.1 | - | 6 | 1457 | 69 | ||
More Information
Set Membership: |
|

MAVS
Genomic Map