Assay Details
| Gene Symbol: ACKR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097430.1 | NP_001090899.1 | 2 | 1053 | 74 | |
| GenBank mRNA | AK230513.1 | - | 4 | 1354 | 74 | |
| AK231126.1 | - | 4 | 1535 | 74 | ||
| AY609393.1 | - | 1 | 997 | 74 | ||
| EF363130.1 | - | 2 | 1053 | 74 | ||
More Information
Set Membership: |
|

ACKR4
Genomic Map