Assay Details
| Gene Symbol: KDM5C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097433.1 | NP_001090902.1 | 24 | 4545 | 115 | |
| XM_005673628.3 | XP_005673685.1 | 4462 | 115 | |||
| XM_005673629.3 | XP_005673686.1 | 4638 | 115 | |||
| XM_005673631.3 | XP_005673688.1 | 4468 | 115 | |||
| XM_005673632.3 | XP_005673689.1 | 4670 | 115 | |||
| XM_013986145.2 | XP_013841599.1 | 4675 | 115 | |||
| XM_013986146.2 | XP_013841600.1 | 4673 | 115 | |||
| XM_013986147.2 | XP_013841601.1 | 4557 | 115 | |||
| XM_013986148.2 | XP_013841602.1 | 4341 | 115 | |||
| XM_013986149.2 | XP_013841603.1 | 4333 | 115 | |||
| XM_021079363.1 | XP_020935022.1 | 4452 | 115 | |||
| XM_021079364.1 | XP_020935023.1 | 4335 | 115 | |||
| GenBank mRNA | EF139241.1 | - | 24 | 4545 | 115 | |
More Information
Set Membership: |
|

KDM5C
Genomic Map