Assay Details
| Gene Symbol: MFSD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097457.1 | NP_001090926.1 | 6 | 2252 | 79 | |
| XM_021074490.1 | XP_020930149.1 | 2637 | 79 | |||
| XM_021074491.1 | XP_020930150.1 | 2306 | 79 | |||
| XM_021074492.1 | XP_020930151.1 | 2562 | 79 | |||
| XM_021074493.1 | XP_020930152.1 | 2659 | 104 | |||
| GenBank mRNA | EF065517.1 | - | 6 | 2252 | 79 | |
More Information
Set Membership: |
|

MFSD6
Genomic Map