Assay Details
| Gene Symbol: CTSB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097458.1 | NP_001090927.1 | 10 | 1053 | 95 | |
| XM_005657265.3 | XP_005657322.3 | 1272 | 95 | |||
| XM_005657266.3 | XP_005657323.1 | 1526 | 95 | |||
| XM_005657267.3 | XP_005657324.1 | 1202 | 95 | |||
| XM_013990293.2 | XP_013845747.1 | 1048 | 95 | |||
| XM_021071763.1 | XP_020927422.1 | 1342 | 95 | |||
| XM_021071764.1 | XP_020927423.1 | 1274 | 95 | |||
| XM_021071765.1 | XP_020927424.1 | 583 | 95 | |||
| GenBank mRNA | AK232389.1 | - | 10 | 1098 | 95 | |
| AK234658.1 | - | 10 | 1042 | 95 | ||
| AK393125.1 | - | 11 | 1213 | 95 | ||
| AK393731.1 | - | 10 | 1065 | 95 | ||
| AK396161.1 | - | 10 | 1098 | 95 | ||
| AK397207.1 | - | 11 | 1235 | 95 | ||
| EF095956.1 | - | 10 | 1053 | 95 | ||
More Information
Set Membership: |
|

CTSB
Genomic Map