Assay Details
| Gene Symbol: HSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097485.1 | NP_001090954.1 | 2 | 143 | 93 | |
| GenBank mRNA | DQ629179.1 | - | 2 | 143 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: HSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097485.1 | NP_001090954.1 | 2 | 143 | 93 | |
| GenBank mRNA | DQ629179.1 | - | 2 | 143 | 93 | |
Set Membership: |
|