Assay Details
| Gene Symbol: ATP5IF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097486.1 | NP_001090955.1 | 3 | 243 | 79 | |
| GenBank mRNA | AK345068.1 | - | 3 | 216 | 79 | |
| AK348940.1 | - | 3 | 216 | 79 | ||
| AK350445.1 | - | 3 | 243 | 79 | ||
| AK400753.1 | - | 3 | 297 | 79 | ||
| DQ629180.1 | - | 3 | 243 | 79 | ||
More Information
Set Membership: |
|

ATP5IF1
Genomic Map