Assay Details
| Gene Symbol: KLF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097487.1 | NP_001090956.1 | 3 | 950 | 113 | |
| XM_021074495.1 | XP_020930154.1 | 1223 | 116 | |||
| XM_021074496.1 | XP_020930155.1 | 1020 | 116 | |||
| XM_021074497.1 | XP_020930156.1 | 1021 | 116 | |||
| GenBank mRNA | AK237548.1 | - | 3 | 996 | 113 | |
| DQ665828.1 | - | 3 | 950 | 113 | ||
More Information
Set Membership: |
|

KLF7
Genomic Map