Assay Details
| Gene Symbol: WEE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097507.2 | NP_001090976.2 | 10 | 1550 | 94 | |
| XM_021078406.1 | XP_020934065.1 | 2338 | 94 | |||
| GenBank mRNA | AB276373.2 | - | 10 | 1550 | 94 | |
More Information
Set Membership: |
|

WEE2
Genomic Map