Assay Details
| Gene Symbol: ADSS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097509.1 | NP_001090978.1 | 1350 | 68 | ||
| XM_021081496.1 | XP_020937155.1 | 1227 | 68 | |||
| XM_021081497.1 | XP_020937156.1 | 1211 | 68 | |||
| GenBank mRNA | AK392100.1 | - | 1373 | 68 | ||
| DQ462751.1 | - | 1350 | 68 | |||
More Information
Set Membership: |
|

ADSS1
Genomic Map