Assay Details
| Gene Symbol: RNASEL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097512.1 | NP_001090981.1 | 7 | 2351 | 129 | |
| GenBank mRNA | DQ497160.1 | - | 7 | 2351 | 129 | |
More Information
Set Membership: |
|

RNASEL
Genomic Map