Assay Details
| Gene Symbol: ADAM3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097513.1 | NP_001090982.1 | 17 | 1849 | 67 | |
| GenBank mRNA | EF525544.1 | - | 17 | 1849 | 67 | |
More Information
Set Membership: |
|

ADAM3A
Genomic Map