Assay Details
| Gene Symbol: TNNI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098599.1 | NP_001092069.1 | 7 | 451 | 70 | |
| XM_021093180.1 | XP_020948839.1 | 2628 | 70 | |||
| XM_021093181.1 | XP_020948840.1 | 5126 | 70 | |||
| XM_021093183.1 | XP_020948842.1 | 3935 | 70 | |||
| XM_021093184.1 | XP_020948843.1 | 4153 | 70 | |||
| GenBank mRNA | AK391776.1 | - | 7 | 527 | 70 | |
| DQ534449.1 | - | 7 | 451 | 70 | ||
| FJ719790.1 | - | 7 | 518 | 70 | ||
More Information
Set Membership: |
|

TNNI3
Genomic Map