Assay Details
| Gene Symbol: SFXN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098602.2 | NP_001092072.1 | 7 | 746 | 61 | |
| XM_005654975.3 | XP_005655032.1 | 913 | 61 | |||
| XM_005654976.3 | XP_005655033.1 | 884 | 61 | |||
| XM_005654978.3 | XP_005655035.1 | 820 | 61 | |||
| XM_013987556.2 | XP_013843010.1 | 818 | 61 | |||
| XM_021076502.1 | XP_020932161.1 | 1008 | 61 | |||
| XM_021076511.1 | XP_020932170.1 | 1304 | 61 | |||
| XM_021076513.1 | XP_020932172.1 | 916 | 61 | |||
| XM_021076518.1 | XP_020932177.1 | 915 | 61 | |||
| GenBank mRNA | AB271931.1 | - | 7 | 746 | 61 | |
| AK343550.1 | - | 7 | 819 | 61 | ||
More Information
Set Membership: |
|

SFXN1
Genomic Map