Assay Details
| Gene Symbol: CALCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104955.1 | NP_001098425.1 | 4 | 353 | 78 | |
| XM_005661110.3 | XP_005661167.1 | 418 | 78 | |||
| GenBank mRNA | AB331639.1 | - | 4 | 353 | 78 | |
More Information
Set Membership: |
|

CALCB
Genomic Map