Assay Details
| Gene Symbol: TRAF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105286.1 | NP_001098756.1 | 6 | 1453 | 79 | |
| XM_005652801.2 | XP_005652858.1 | 1739 | 79 | |||
| XM_013990069.2 | XP_013845523.1 | 1617 | 79 | |||
| GenBank mRNA | AK393349.1 | - | 7 | 1684 | 79 | |
| EU095967.1 | - | 6 | 1453 | 79 | ||
More Information
Set Membership: |
|

TRAF6
Genomic Map