Assay Details
| Gene Symbol: CD302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110425.1 | NP_001103895.1 | 6 | 652 | 108 | |
| GenBank mRNA | AK233610.1 | - | 6 | 687 | 108 | |
| AK398679.1 | - | 6 | 108 | |||
| AY609741.1 | - | 5 | 542 | 108 | ||
More Information
Set Membership: |
|

CD302
Genomic Map