Assay Details
| Gene Symbol: SUMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112676.1 | NP_001106146.1 | 1 | 146 | 127 | |
| GenBank mRNA | AB355636.1 | - | 3 | 231 | 127 | |
| AK238892.1 | - | 3 | 280 | 127 | ||
| AK343895.1 | - | 3 | 258 | 127 | ||
| AK392044.1 | - | 3 | 259 | 127 | ||
| AM397625.1 | - | 1 | 146 | 127 | ||
| AY609886.1 | - | 3 | 271 | 127 | ||
| DQ886441.1 | - | 1 | 146 | 127 | ||
| Gene Symbol: LOC100737265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005659490.2 | XP_005659547.1 | 173 | 127 | ||
More Information
Set Membership: |
|

SUMO1
Genomic Map