Assay Details
| Gene Symbol: HNRNPAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001112695.1 | NP_001106165.1 | 4 | 653 | 142 | |
| GenBank mRNA | AB304402.1 | - | 4 | 653 | 142 | |
| AK237336.1 | - | 5 | 881 | 142 | ||
| AK396943.1 | - | 5 | 887 | 142 | ||
| AK398859.1 | - | 5 | 876 | 142 | ||
More Information
Set Membership: |
|

HNRNPAB
Genomic Map