Assay Details
| Gene Symbol: FADS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113041.1 | NP_001106512.1 | 11 | 1250 | 81 | |
| GenBank mRNA | AB933145.1 | - | 11 | 1256 | 81 | |
| AB933146.1 | - | 11 | 1130 | 81 | ||
| AK394022.1 | - | 9 | 850 | 81 | ||
| EU131180.1 | - | 11 | 1250 | 81 | ||
More Information
Set Membership: |
|

FADS1
Genomic Map