Assay Details
| Gene Symbol: TAS1R3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113288.1 | NP_001106759.1 | 5 | 1854 | 72 | |
| GenBank mRNA | AB162127.1 | - | 5 | 1854 | 72 | |
| AB162128.1 | - | 5 | 1804 | 72 | ||
| AB162129.1 | - | 6 | 1730 | 72 | ||
More Information
Set Membership: |
|

TAS1R3
Genomic Map