Assay Details
Gene Symbol: KEAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114671.1 | NP_001108143.1 | 6 | 1951 | 104 | |
XM_005654811.3 | XP_005654868.1 | 2296 | 104 | |||
XM_013987396.2 | XP_013842850.1 | 1970 | 104 | |||
XM_021076654.1 | XP_020932313.1 | 2128 | 104 | |||
XM_021076667.1 | XP_020932326.1 | 1828 | 104 | |||
GenBank mRNA | AK231409.1 | - | 6 | 1860 | 104 | |
AK393862.1 | - | 4 | 593 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
