Assay Details
| Gene Symbol: GPX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115136.1 | NP_001108608.1 | 2 | 650 | 100 | |
| GenBank mRNA | AK345250.1 | - | 2 | 646 | 100 | |
| AK392829.1 | - | 2 | 649 | 100 | ||
| CJ026838.1 | - | 2 | 650 | 100 | ||
| DQ898282.2 | - | 2 | 650 | 100 | ||
More Information
Set Membership: |
|

GPX2
Genomic Map