Assay Details
| Gene Symbol: ADRA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123072.1 | NP_001116544.1 | 2 | 1251 | 86 | |
| XM_005657244.3 | XP_005657301.1 | 2513 | 86 | |||
| XM_005657245.3 | XP_005657302.1 | 2061 | 86 | |||
| XM_005657246.3 | XP_005657303.1 | 2590 | 86 | |||
| XM_013990269.2 | XP_013845723.1 | 2383 | 86 | |||
| XM_021071783.1 | XP_020927442.1 | 1920 | 86 | |||
More Information
Set Membership: |
|

ADRA1A
Genomic Map