Assay Details
| Gene Symbol: ADRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123074.1 | NP_001116546.1 | 1 | 649 | 89 | |
| GenBank mRNA | U56425.1 | - | 1 | 353 | 89 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ADRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123074.1 | NP_001116546.1 | 1 | 649 | 89 | |
| GenBank mRNA | U56425.1 | - | 1 | 353 | 89 | |
Set Membership: |
|