Assay Details
| Gene Symbol: DRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123108.1 | NP_001116580.1 | 2 | 2154 | 138 | |
| XM_021076520.1 | XP_020932179.1 | 138 | ||||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: DRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123108.1 | NP_001116580.1 | 2 | 2154 | 138 | |
| XM_021076520.1 | XP_020932179.1 | 138 | ||||
Set Membership: |
|