Assay Details
| Gene Symbol: HPCA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123125.1 | NP_001116597.1 | 2 | 445 | 114 | |
| XM_005665170.3 | XP_005665227.1 | 546 | 114 | |||
| XM_005665171.3 | XP_005665228.2 | 714 | 114 | |||
| GenBank mRNA | DQ917644.1 | - | 2 | 445 | 114 | |
More Information
Set Membership: |
|

HPCA
Genomic Map