Assay Details
| Gene Symbol: LIN28A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123133.1 | NP_001116605.1 | 2 | 153 | 61 | |
| GenBank mRNA | EU503118.1 | - | 2 | 153 | 61 | |
| HM347046.1 | - | 2 | 153 | 61 | ||
More Information
Set Membership: |
|

LIN28A
Genomic Map