Assay Details
| Gene Symbol: RAP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123185.1 | NP_001116657.1 | 1 | 455 | 60 | |
| GenBank mRNA | DQ917633.1 | - | 1 | 455 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RAP2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123185.1 | NP_001116657.1 | 1 | 455 | 60 | |
| GenBank mRNA | DQ917633.1 | - | 1 | 455 | 60 | |
Set Membership: |
|