Assay Details
| Gene Symbol: TPPP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123206.1 | NP_001116678.1 | 4 | 549 | 59 | |
| XM_021076519.1 | XP_020932178.1 | 619 | 59 | |||
| GenBank mRNA | EU522721.1 | - | 4 | 549 | 59 | |
More Information
Set Membership: |
|

TPPP
Genomic Map