Assay Details
| Gene Symbol: GGTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323055.1 | NP_001309984.1 | 6 | 1470 | 84 | |
| NM_001323056.1 | NP_001309985.1 | 7 | 1582 | 84 | ||
| NM_213810.3 | NP_998975.1 | 8 | 1618 | 84 | ||
| XM_005660399.3 | XP_005660456.1 | 1294 | 84 | |||
| XM_005660402.3 | XP_005660459.1 | 981 | 84 | |||
| XM_013993814.2 | XP_013849268.1 | 1086 | 84 | |||
| XM_013993815.2 | XP_013849269.1 | 969 | 84 | |||
| XM_013993817.2 | XP_013849271.1 | 864 | 84 | |||
| XM_021081683.1 | XP_020937342.1 | 1072 | 84 | |||
| XM_021081694.1 | XP_020937353.1 | 1189 | 84 | |||
| GenBank mRNA | AF221508.1 | - | 8 | 1621 | 84 | |
| AF221509.1 | - | 7 | 1577 | 84 | ||
| AK234328.1 | - | 7 | 1290 | 84 | ||
| AK347936.1 | - | 6 | 1086 | 84 | ||
| AK400512.1 | - | 6 | 1256 | 84 | ||
| L36152.1 | - | 6 | 961 | 84 | ||
| L36535.1 | - | 6 | 959 | 84 | ||
More Information
Set Membership: |
|

GGTA1
Genomic Map