Assay Details
| Gene Symbol: QDPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214343.2 | NP_999508.1 | 7 | 648 | 73 | |
| GenBank mRNA | AF526879.2 | - | 7 | 648 | 73 | |
| AY609903.1 | - | 7 | 656 | 73 | ||
More Information
Set Membership: |
|

QDPR
Genomic Map