Assay Details
| Gene Symbol: SRY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214452.3 | NP_999617.2 | 1 | 553 | 65 | |
| Gene Symbol: LOC106504154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013986578.2 | XP_013842032.1 | 613 | 65 | ||
More Information
Set Membership: |
|

SRY
Genomic Map