Assay Details
| Gene Symbol: RPSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037146.2 | NP_001032223.1 | 5 - 6 | 679 | 78 | |
| GenBank mRNA | AK231521.1 | - | 5 - 6 | 698 | 78 | |
| AK233707.1 | - | 5 - 6 | 698 | 78 | ||
| AK236883.1 | - | 5 - 6 | 765 | 78 | ||
| AK239730.1 | - | 5 - 6 | 700 | 78 | ||
| AK240295.1 | - | 5 - 6 | 702 | 78 | ||
| AK345661.1 | - | 5 - 6 | 698 | 78 | ||
| AK346582.1 | - | 5 - 6 | 714 | 78 | ||
| AK346667.1 | - | 5 - 6 | 699 | 78 | ||
| AK352307.1 | - | 5 - 6 | 698 | 78 | ||
| AK392740.1 | - | 5 - 6 | 699 | 78 | ||
| AK392954.1 | - | 5 - 6 | 704 | 78 | ||
| AK394403.1 | - | 5 - 6 | 705 | 78 | ||
| AK398657.1 | - | 5 - 6 | 698 | 78 | ||
| AK398723.1 | - | 5 - 6 | 701 | 78 | ||
| AK398868.1 | - | 5 - 6 | 703 | 78 | ||
| AK398981.1 | - | 5 - 6 | 813 | 78 | ||
| AK398991.1 | - | 5 - 6 | 705 | 78 | ||
| AK398995.1 | - | 5 - 6 | 699 | 78 | ||
| AK399002.1 | - | 5 - 6 | 706 | 78 | ||
| AK400775.1 | - | 5 - 6 | 699 | 78 | ||
| AK400937.1 | - | 5 - 6 | 704 | 78 | ||
| AM050292.2 | - | 5 - 6 | 706 | 78 | ||
More Information
Set Membership: |
|

RPSA
Genomic Map