Assay Details
| Gene Symbol: RPSA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037146.2 | NP_001032223.1 | 6 - 7 | 847 | 78 | |
| GenBank mRNA | AK231521.1 | - | 6 - 7 | 866 | 78 | |
| AK236825.1 | - | 6 - 7 | 867 | 78 | ||
| AK236883.1 | - | 6 - 7 | 933 | 78 | ||
| AK239730.1 | - | 6 - 7 | 868 | 78 | ||
| AK240295.1 | - | 6 - 7 | 870 | 78 | ||
| AK345661.1 | - | 6 - 7 | 866 | 78 | ||
| AK346582.1 | - | 6 - 7 | 882 | 78 | ||
| AK346667.1 | - | 6 - 7 | 867 | 78 | ||
| AK352307.1 | - | 6 - 7 | 866 | 78 | ||
| AK392740.1 | - | 6 - 7 | 867 | 78 | ||
| AK392954.1 | - | 6 - 7 | 872 | 78 | ||
| AK394403.1 | - | 6 - 7 | 873 | 78 | ||
| AK398657.1 | - | 6 - 7 | 866 | 78 | ||
| AK398723.1 | - | 6 - 7 | 869 | 78 | ||
| AK398868.1 | - | 6 - 7 | 871 | 78 | ||
| AK398981.1 | - | 6 - 7 | 981 | 78 | ||
| AK398991.1 | - | 6 - 7 | 873 | 78 | ||
| AK398995.1 | - | 6 - 7 | 867 | 78 | ||
| AK399002.1 | - | 6 - 7 | 874 | 78 | ||
| AK400775.1 | - | 6 - 7 | 867 | 78 | ||
| AK400937.1 | - | 6 - 7 | 872 | 78 | ||
| AM050292.2 | - | 6 - 7 | 874 | 78 | ||
More Information
Set Membership: |
|

RPSA
Genomic Map