Assay Details
| Gene Symbol: PPP1CA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044559.1 | NP_001038024.1 | 3 - 4 | 471 | 66 | |
| GenBank mRNA | AK345871.1 | - | 3 - 4 | 508 | 66 | |
| AK391575.1 | - | 3 - 4 | 513 | 66 | ||
| DQ380140.1 | - | 3 - 4 | 471 | 66 | ||
More Information
Set Membership: |
|

PPP1CA
Genomic Map