Assay Details
| Gene Symbol: PPP1CC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044560.1 | NP_001038025.1 | 6 - 7 | 877 | 80 | |
| GenBank mRNA | AK235498.1 | - | 6 - 7 | 1109 | 80 | |
| AK392314.1 | - | 6 - 7 | 1099 | 80 | ||
| DQ380141.1 | - | 6 - 7 | 877 | 80 | ||
More Information
Set Membership: |
|

PPP1CC
Genomic Map