Assay Details
| Gene Symbol: RHCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044577.1 | NP_001038042.1 | 9 - 10 | 1356 | 62 | |
| XM_021098368.1 | XP_020954027.1 | 1382 | 62 | |||
| XM_021098369.1 | XP_020954028.1 | 1382 | 62 | |||
| GenBank mRNA | AK236155.1 | - | 9 - 10 | 1353 | 62 | |
| AK391229.1 | - | 9 - 10 | 1386 | 62 | ||
| DQ523515.1 | - | 9 - 10 | 1356 | 62 | ||
More Information
Set Membership: |
|

RHCG
Genomic Map