Assay Details
| Gene Symbol: TAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044581.1 | NP_001038046.1 | 5 - 6 | 1260 | 91 | |
| XM_021098370.1 | XP_020954029.1 | 1429 | 91 | |||
| GenBank mRNA | AK390391.1 | - | 5 - 6 | 1505 | 91 | |
| DQ227989.1 | - | 5 - 6 | 1260 | 91 | ||
| DQ227990.1 | - | 5 - 6 | 1233 | 91 | ||
More Information
Set Membership: |
|

TAP1
Genomic Map