Assay Details
| Gene Symbol: FBXO32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044588.1 | NP_001038053.1 | 4 - 5 | 414 | 87 | |
| GenBank mRNA | AK231687.1 | - | 4 - 5 | 656 | 87 | |
| AK235526.1 | - | 4 - 5 | 978 | 87 | ||
| AK393264.1 | - | 4 - 5 | 599 | 87 | ||
| AK396556.1 | - | 4 - 5 | 602 | 87 | ||
| AK400295.1 | - | 1 - 2 | 748 | 87 | ||
| AK400532.1 | - | 4 - 5 | 653 | 87 | ||
| AY610094.1 | - | 4 - 5 | 587 | 87 | ||
| DQ288235.1 | - | 4 - 5 | 414 | 87 | ||
More Information
Set Membership: |
|

FBXO32
Genomic Map