Assay Details
| Gene Symbol: CST3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044602.1 | NP_001038067.1 | 1 - 2 | 264 | 99 | |
| GenBank mRNA | AK392782.1 | - | 1 - 2 | 299 | 99 | |
| AK399954.1 | - | 1 - 2 | 322 | 99 | ||
| DQ661047.1 | - | 1 - 2 | 264 | 99 | ||
More Information
Set Membership: |
|

CST3
Genomic Map