Assay Details
| Gene Symbol: TUBB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044612.1 | NP_001038077.1 | 2 - 3 | 373 | 64 | |
| GenBank mRNA | AK238600.1 | - | 2 - 3 | 466 | 64 | |
| AK390248.1 | - | 1 - 2 | 486 | 64 | ||
| AK392245.1 | - | 2 - 3 | 342 | 64 | ||
| AK400507.1 | - | 2 - 3 | 348 | 64 | ||
| DQ225361.1 | - | 2 - 3 | 373 | 64 | ||
More Information
Set Membership: |
|

TUBB
Genomic Map