Assay Details
| Gene Symbol: RIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077230.2 | NP_001070698.2 | 5 - 6 | 608 | 65 | |
| XM_021088570.1 | XP_020944229.1 | 474 | 65 | |||
| GenBank mRNA | AK344536.1 | - | 5 - 6 | 607 | 65 | |
| AY609626.1 | - | 4 - 5 | 477 | 65 | ||
| DQ972971.1 | - | 4 - 5 | 431 | 65 | ||
More Information
Set Membership: |
|

RIT1
Genomic Map