Assay Details
| Gene Symbol: CPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097439.1 | NP_001090908.1 | 5 - 6 | 972 | 60 | |
| GenBank mRNA | AK392225.1 | - | 5 - 6 | 1126 | 60 | |
| AM042539.1 | - | 5 - 6 | 972 | 60 | ||
More Information
Set Membership: |
|

CPE
Genomic Map