Assay Details
Gene Symbol: C8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001097450.2 | NP_001090919.2 | 3 - 4 | 426 | 75 | |
XM_021093175.1 | XP_020948834.1 | 432 | 75 | |||
GenBank mRNA | AB008156.1 | - | 3 - 4 | 414 | 75 | |
AK232357.1 | - | 3 - 4 | 426 | 75 | ||
DQ333200.1 | - | 3 - 4 | 414 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
