Assay Details
| Gene Symbol: CAPZA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097454.1 | NP_001090923.1 | 5 - 6 | 424 | 81 | |
| GenBank mRNA | AK233290.1 | - | 5 - 6 | 1014 | 81 | |
| AK389793.1 | - | 5 - 6 | 437 | 81 | ||
| AK391682.1 | - | 5 - 6 | 438 | 81 | ||
| AM410994.1 | - | 5 - 6 | 424 | 81 | ||
| EF202988.1 | - | 5 - 6 | 461 | 81 | ||
More Information
Set Membership: |
|

CAPZA1
Genomic Map