Assay Details
| Gene Symbol: RPL10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097477.2 | NP_001090946.2 | 3 - 4 | 216 | 90 | |
| XM_021097852.1 | XP_020953511.1 | 180 | 90 | |||
| GenBank mRNA | AK240001.1 | - | 3 - 4 | 191 | 90 | |
| AK347871.1 | - | 3 - 4 | 186 | 90 | ||
| AK350326.1 | - | 812 | 90 | |||
| AK392651.1 | - | 3 - 4 | 185 | 90 | ||
| DB788284.1 | - | 3 - 4 | 184 | 90 | ||
| DQ629164.1 | - | 3 - 4 | 182 | 91 | ||
| HX212267.1 | - | 3 - 4 | 217 | 90 | ||
More Information
Set Membership: |
|

RPL10A
Genomic Map